Assay Details
| Gene Symbol: tmem205 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080686.1 | NP_001074155.1 | 4 | 1551 | 139 | |
| XM_005168478.3 | XP_005168535.1 | 4 | 1615 | 139 | ||
| GenBank mRNA | BC129498.1 | - | 4 | 1551 | 139 | |
More Information
Set Membership: |
|

tmem205
Genomic Map