Assay Details
| Gene Symbol: htra1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002219.1 | NP_001002219.1 | 1 | 353 | 114 | |
| GenBank mRNA | BC074069.1 | - | 1 | 353 | 114 | |
| BC165751.1 | - | 1 | 249 | 114 | ||
More Information
Set Membership: |
|

htra1a
Genomic Map