Assay Details
| Gene Symbol: psmc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199750.2 | NP_956044.2 | 11 | 1500 | 137 | |
| GenBank mRNA | BC055215.1 | - | 11 | 1449 | 137 | |
| BC153479.1 | - | 11 | 1442 | 137 | ||
More Information
Set Membership: |
|

psmc4
Genomic Map