Assay Details
| Gene Symbol: abhd11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004290.2 | NP_001004290.1 | 6 | 1057 | 111 | |
| XM_009295304.3 | XP_009293579.1 | 6 | 1063 | 111 | ||
| GenBank mRNA | BC134019.1 | - | 6 | 1057 | 111 | |
More Information
Set Membership: |
|

abhd11
Genomic Map