Assay Details
| Gene Symbol: nap1l4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001089347.1 | NP_001082816.1 | 15 | 1851 | 120 | |
| XM_005170675.4 | XP_005170732.1 | 14 | 1812 | 120 | ||
| GenBank mRNA | BC139678.1 | - | 15 | 1851 | 120 | |
More Information
Set Membership: |
|

nap1l4a
Genomic Map