Assay Details
| Gene Symbol: nefmb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123280.1 | NP_001116752.1 | 3 | 2577 | 148 | |
| GenBank mRNA | BC133105.1 | - | 4 | 2547 | 148 | |
More Information
Set Membership: |
|

nefmb
Genomic Map