Assay Details
| Gene Symbol: trmt11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200216.2 | NP_956510.2 | 11 | 1351 | 84 | |
| XM_021466641.1 | XP_021322316.1 | 11 | 1114 | 84 | ||
| GenBank mRNA | AF532981.1 | - | 11 | 1351 | 84 | |
| BC046012.1 | - | 11 | 1093 | 84 | ||
More Information
Set Membership: |
|

trmt11
Genomic Map