Assay Details
| Gene Symbol: kat7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076584.1 | NP_001070052.1 | 3 | 1247 | 138 | |
| GenBank mRNA | BC124518.1 | - | 3 | 1247 | 138 | |
More Information
Set Membership: |
|

kat7a
Genomic Map