Assay Details
| Gene Symbol: actn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199586.1 | NP_955880.1 | 21 | 3947 | 96 | |
| XM_021481334.1 | XP_021337009.1 | 22 | 4041 | 96 | ||
| XM_021481335.1 | XP_021337010.1 | 22 | 4041 | 96 | ||
| XM_021481336.1 | XP_021337011.1 | 22 | 4041 | 96 | ||
| XM_021481337.1 | XP_021337012.1 | 21 | 3975 | 96 | ||
| XM_021481338.1 | XP_021337013.1 | 21 | 3960 | 96 | ||
| XM_021481339.1 | XP_021337014.1 | 21 | 3960 | 96 | ||
| GenBank mRNA | BC054901.1 | - | 21 | 3947 | 96 | |
More Information
Set Membership: |
|

actn4
Genomic Map