Assay Details
| Gene Symbol: znf148 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099234.1 | NP_001092704.1 | 8 | 1256 | 127 | |
| GenBank mRNA | BC142733.1 | - | 8 | 1256 | 127 | |
More Information
Set Membership: |
|

znf148
Genomic Map