Assay Details
| Gene Symbol: pmvk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083579.1 | NP_001077048.1 | 5 | 749 | 145 | |
| GenBank mRNA | BC135045.1 | - | 5 | 749 | 145 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

pmvk| Gene Symbol: pmvk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083579.1 | NP_001077048.1 | 5 | 749 | 145 | |
| GenBank mRNA | BC135045.1 | - | 5 | 749 | 145 | |
Set Membership: |
|