Assay Details
| Gene Symbol: ccdc127b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082883.1 | NP_001076352.1 | 3 | 1051 | 115 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ccdc127b| Gene Symbol: ccdc127b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082883.1 | NP_001076352.1 | 3 | 1051 | 115 | |
Set Membership: |
|