Assay Details
| Gene Symbol: lhpp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098781.1 | NP_001092251.1 | 7 | 852 | 155 | |
| GenBank mRNA | BC142939.1 | - | 7 | 852 | 155 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

lhpp| Gene Symbol: lhpp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098781.1 | NP_001092251.1 | 7 | 852 | 155 | |
| GenBank mRNA | BC142939.1 | - | 7 | 852 | 155 | |
Set Membership: |
|