Assay Details
| Gene Symbol: cpe | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214810.2 | NP_999975.1 | 1 | 248 | 100 | |
| GenBank mRNA | BC070026.1 | - | 1 | 248 | 100 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cpe| Gene Symbol: cpe | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214810.2 | NP_999975.1 | 1 | 248 | 100 | |
| GenBank mRNA | BC070026.1 | - | 1 | 248 | 100 | |
Set Membership: |
|