Assay Details
| Gene Symbol: nrl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC163220.1 | - | 1 | 490 | 146 | |
| DQ018734.1 | - | 1 | 451 | 146 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

nrl| Gene Symbol: nrl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC163220.1 | - | 1 | 490 | 146 | |
| DQ018734.1 | - | 1 | 451 | 146 | ||
Set Membership: |
|