Assay Details
| Gene Symbol: etv7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083024.1 | NP_001076493.1 | 9 | 1649 | 82 | |
| GenBank mRNA | BC133152.1 | - | 9 | 1649 | 82 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

etv7| Gene Symbol: etv7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083024.1 | NP_001076493.1 | 9 | 1649 | 82 | |
| GenBank mRNA | BC133152.1 | - | 9 | 1649 | 82 | |
Set Membership: |
|