Assay Details
| Gene Symbol: dnajb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003455.1 | NP_001003455.1 | 2 | 850 | 102 | |
| GenBank mRNA | BC077166.1 | - | 2 | 850 | 102 | |
| BC164144.1 | - | 2 | 704 | 102 | ||
More Information
Set Membership: |
|

dnajb4
Genomic Map