Assay Details
| Gene Symbol: cavin2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004584.1 | NP_001004584.2 | 2 | 2741 | 120 | |
| GenBank mRNA | BC081627.1 | - | 2 | 2741 | 120 | |
More Information
Set Membership: |
|

cavin2b
Genomic Map