Assay Details
| Gene Symbol: rdh12l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009912.1 | NP_001009912.1 | 5 | 946 | 74 | |
| GenBank mRNA | BC078208.1 | - | 5 | 946 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

rdh12l| Gene Symbol: rdh12l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009912.1 | NP_001009912.1 | 5 | 946 | 74 | |
| GenBank mRNA | BC078208.1 | - | 5 | 946 | 74 | |
Set Membership: |
|