Assay Details
| Gene Symbol: gna13a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012243.3 | NP_001012243.3 | 4 | 1185 | 116 | |
| GenBank mRNA | BC095814.1 | - | 4 | 1152 | 116 | |
| EH567709.1 | - | 1 | 208 | 116 | ||
More Information
Set Membership: |
|

gna13a
Genomic Map