Assay Details
| Gene Symbol: ext1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012369.2 | NP_001012369.2 | 11 | 2750 | 71 | |
| GenBank mRNA | BC125900.1 | - | 11 | 2750 | 71 | |
More Information
Set Membership: |
|

ext1b
Genomic Map