Assay Details
| Gene Symbol: tmem222a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013316.3 | NP_001013334.2 | 6 | 2934 | 138 | |
| GenBank mRNA | BC129140.1 | - | 6 | 2850 | 138 | |
More Information
Set Membership: |
|

tmem222a
Genomic Map