Assay Details
| Gene Symbol: ctbp2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015064.1 | 757 | 67 | |||
| GenBank mRNA | AY878351.1 | - | 757 | 67 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ctbp2l| Gene Symbol: ctbp2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015064.1 | 757 | 67 | |||
| GenBank mRNA | AY878351.1 | - | 757 | 67 | ||
Set Membership: |
|