Assay Details
| Gene Symbol: b4galt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017730.1 | NP_001017730.1 | 3 | 1053 | 114 | |
| GenBank mRNA | BC093107.1 | - | 3 | 1053 | 114 | |
More Information
Set Membership: |
|

b4galt1
Genomic Map