Assay Details
| Gene Symbol: dusp22b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017742.2 | NP_001017742.2 | 7 | 1052 | 129 | |
| GenBank mRNA | BC093370.1 | - | 7 | 1044 | 129 | |
More Information
Set Membership: |
|

dusp22b
Genomic Map