Assay Details
| Gene Symbol: cryba4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001018125.1 | NP_001018135.1 | 5 | 451 | 68 | |
| XM_009305688.3 | XP_009303963.1 | 5 | 504 | 68 | ||
| GenBank mRNA | BC095102.1 | - | 5 | 451 | 68 | |
| BC165263.1 | - | 4 | 442 | 68 | ||
| DQ000464.1 | - | 5 | 442 | 68 | ||
More Information
Set Membership: |
|

cryba4
Genomic Map