Assay Details
| Gene Symbol: hs6st1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020508.2 | NP_001018344.2 | 2 | 1547 | 83 | |
| GenBank mRNA | BC095149.1 | - | 2 | 1547 | 83 | |
More Information
Set Membership: |
|

hs6st1a
Genomic Map