Assay Details
| Gene Symbol: ptk7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020665.1 | NP_001018501.1 | 1 | 146 | 107 | |
| GenBank mRNA | BC095690.1 | - | 1 | 146 | 107 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC095690.1, NM_001020665.1 transcripts.
Set Membership: |
|

ptk7a
Genomic Map