Assay Details
| Gene Symbol: bloc1s5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030066.1 | NP_001025237.1 | 5 | 1149 | 68 | |
| GenBank mRNA | BC096921.1 | - | 5 | 1149 | 68 | |
| BC153651.1 | - | 5 | 1104 | 68 | ||
More Information
Set Membership: |
|

bloc1s5
Genomic Map