Assay Details
| Gene Symbol: aqp9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033096.1 | NP_001028268.1 | 1456 | 140 | ||
| GenBank mRNA | BC100051.1 | - | 1456 | 140 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

aqp9a| Gene Symbol: aqp9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033096.1 | NP_001028268.1 | 1456 | 140 | ||
| GenBank mRNA | BC100051.1 | - | 1456 | 140 | ||
Set Membership: |
|