Assay Details
| Gene Symbol: pbxip1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079956.2 | NP_001073425.2 | 9 | 5078 | 101 | |
| XM_017351348.2 | XP_017206837.1 | 11 | 5017 | 101 | ||
| GenBank mRNA | BC129142.1 | - | 9 | 5051 | 101 | |
More Information
Set Membership: |
|

pbxip1b
Genomic Map