Assay Details
| Gene Symbol: dnlz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080648.1 | NP_001074117.1 | 3 | 951 | 146 | |
| GenBank mRNA | BC129164.1 | - | 3 | 951 | 146 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

dnlz| Gene Symbol: dnlz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080648.1 | NP_001074117.1 | 3 | 951 | 146 | |
| GenBank mRNA | BC129164.1 | - | 3 | 951 | 146 | |
Set Membership: |
|