Assay Details
| Gene Symbol: caspxa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083862.1 | NP_001077331.1 | 1 | 53 | 74 | |
| Gene Symbol: casp20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136252.1 | NP_001129724.1 | 1 | 136 | 74 | |
| XM_009302221.3 | XP_009300496.1 | 2 | 197 | 74 | ||
| GenBank mRNA | BC163666.1 | - | 1 | 136 | 74 | |
| DQ812122.1 | - | 1 | 53 | 74 | ||
| MG958007.1 | - | 1 | 53 | 74 | ||
More Information
Set Membership: |
|

caspxa
Genomic Map