Assay Details
| Gene Symbol: lrrc3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001089516.1 | NP_001082985.1 | 2 | 751 | 92 | |
| XM_005173724.4 | XP_005173781.1 | 2 | 1252 | 92 | ||
| GenBank mRNA | BC133927.1 | - | 2 | 751 | 92 | |
| CU468754.1 | - | 1 | 462 | 92 | ||
More Information
Set Membership: |
|

lrrc3b
Genomic Map