Assay Details
| Gene Symbol: zgc:165573 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098766.1 | NP_001092236.1 | 1 | 54 | 83 | |
| GenBank mRNA | BC091540.1 | - | 1 | 112 | 83 | |
| BC124722.1 | - | 1 | 111 | 83 | ||
| BC142871.1 | - | 1 | 54 | 83 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC142871.1, NM_001098766.1 transcripts.
Set Membership: |
|

zgc:165573
Genomic Map