Assay Details
| Gene Symbol: cd247 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100157.1 | NP_001093627.1 | 46 | 53 | ||
| GenBank mRNA | EF601086.1 | - | 46 | 53 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cd247| Gene Symbol: cd247 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100157.1 | NP_001093627.1 | 46 | 53 | ||
| GenBank mRNA | EF601086.1 | - | 46 | 53 | ||
Set Membership: |
|