Assay Details
| Gene Symbol: pou3f2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131160.1 | NP_571235.1 | 1 | 2853 | 143 | |
| GenBank mRNA | BC067373.1 | - | 1 | 2814 | 144 | |
| Y07905.1 | - | 2853 | 143 | |||
More Information
Set Membership: |
|

pou3f2b
Genomic Map