Assay Details
| Gene Symbol: dbx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021470994.1 | XP_021326669.1 | 487 | 73 | ||
| GenBank mRNA | AF030286.1 | - | 455 | 73 | ||
| BC091853.1 | - | 457 | 73 | |||
| BC165905.1 | - | 483 | 73 | |||
More Information
Set Membership: |
|

dbx2
Genomic Map