Assay Details
| Gene Symbol: snrpd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173252.2 | NP_775359.1 | 4 | 532 | 130 | |
| GenBank mRNA | AF506225.1 | - | 4 | 450 | 130 | |
| BC071543.1 | - | 4 | 488 | 130 | ||
| EH445570.1 | - | 4 | 497 | 130 | ||
| FP210948.1 | - | 4 | 485 | 130 | ||
More Information
Set Membership: |
|

snrpd1
Genomic Map