Assay Details
| Gene Symbol: sept9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198911.1 | NP_944593.1 | 11 | 2753 | 101 | |
| GenBank mRNA | AY398383.1 | - | 11 | 2753 | 101 | |
| BC061455.1 | - | 11 | 2837 | 101 | ||
More Information
Set Membership: |
|

sept9a
Genomic Map