Assay Details
| Gene Symbol: h1-0 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199552.1 | NP_955846.1 | 1 | 952 | 92 | |
| GenBank mRNA | BC056596.1 | - | 1 | 952 | 92 | |
| BC066449.1 | - | 1 | 940 | 92 | ||
More Information
Set Membership: |
|

h1-0
Genomic Map