Assay Details
| Gene Symbol: ppp1cbl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199916.1 | NP_956210.1 | 1046 | 62 | ||
| GenBank mRNA | BC053296.1 | - | 1046 | 62 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ppp1cbl| Gene Symbol: ppp1cbl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199916.1 | NP_956210.1 | 1046 | 62 | ||
| GenBank mRNA | BC053296.1 | - | 1046 | 62 | ||
Set Membership: |
|