Assay Details
| Gene Symbol: tarbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199997.1 | NP_956291.1 | 7 | 1556 | 101 | |
| GenBank mRNA | BC055390.1 | - | 7 | 1556 | 101 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

tarbp2| Gene Symbol: tarbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199997.1 | NP_956291.1 | 7 | 1556 | 101 | |
| GenBank mRNA | BC055390.1 | - | 7 | 1556 | 101 | |
Set Membership: |
|