Assay Details
| Gene Symbol: purbb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200547.1 | NP_956841.1 | 1 | 1555 | 88 | |
| GenBank mRNA | BC056517.1 | - | 1 | 1555 | 88 | |
| BC067670.1 | - | 1 | 1502 | 88 | ||
More Information
Set Membership: |
|

purbb
Genomic Map