Assay Details
| Gene Symbol: desi1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200700.1 | NP_956994.1 | 6 | 1943 | 156 | |
| GenBank mRNA | BC059442.1 | - | 6 | 1943 | 156 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

desi1a| Gene Symbol: desi1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200700.1 | NP_956994.1 | 6 | 1943 | 156 | |
| GenBank mRNA | BC059442.1 | - | 6 | 1943 | 156 | |
Set Membership: |
|