Assay Details
| Gene Symbol: mrpl57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200775.1 | NP_957069.1 | 2 | 348 | 65 | |
| GenBank mRNA | BC059609.1 | - | 2 | 348 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mrpl57| Gene Symbol: mrpl57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200775.1 | NP_957069.1 | 2 | 348 | 65 | |
| GenBank mRNA | BC059609.1 | - | 2 | 348 | 65 | |
Set Membership: |
|