Assay Details
| Gene Symbol: rhcgb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320382.1 | NP_001307311.1 | 1 | 72 | 79 | |
| XM_017354103.2 | XP_017209592.2 | 1 | 92 | 79 | ||
| GenBank mRNA | AB286865.1 | - | 1 | 69 | 79 | |
| AF398238.1 | - | 1 | 50 | 79 | ||
| BC066492.1 | - | 50 | 79 | |||
| EH442849.1 | - | 1 | 96 | 79 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC066492.1, NM_001320382.1, XM_017354103.2 transcripts.
Set Membership: |
|

rhcgb
Genomic Map