Assay Details
| Gene Symbol: junbb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212750.1 | NP_997915.1 | 1 | 1544 | 101 | |
| GenBank mRNA | BC053154.1 | - | 1 | 1544 | 101 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

junbb| Gene Symbol: junbb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212750.1 | NP_997915.1 | 1 | 1544 | 101 | |
| GenBank mRNA | BC053154.1 | - | 1 | 1544 | 101 | |
Set Membership: |
|