Assay Details
| Gene Symbol: mbd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212768.1 | NP_997933.1 | 746 | 57 | ||
| GenBank mRNA | AY238336.1 | - | 746 | 57 | ||
| BC093117.1 | - | 635 | 57 | |||
| BC153555.1 | - | 727 | 57 | |||
| BC164452.1 | - | 498 | 57 | |||
More Information
Set Membership: |
|

mbd2
Genomic Map