Assay Details
| Gene Symbol: serpinb5l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212877.1 | NP_998042.1 | 149 | 71 | ||
| GenBank mRNA | BC066740.1 | - | 149 | 71 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

serpinb5l| Gene Symbol: serpinb5l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212877.1 | NP_998042.1 | 149 | 71 | ||
| GenBank mRNA | BC066740.1 | - | 149 | 71 | ||
Set Membership: |
|